missing translation for 'onlineSavingsMsg'
Learn More

2-(Carboxymethylthio)pyrimidine, 98%, Thermo Scientific™

Catalog Number p-7080919
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
88768-45-0
C6H5N2O2S
169.18
MFCD00010287
NIEOYUNNKKAQKI-UHFFFAOYSA-M
2-pyrimidylthio acetic acid, pyrimidin-2-ylthio acetic acid, 2-pyrimidin-2-ylthio acetic acid, pyrimidin-2-ylsulfanyl acetic acid, 2-pyrimidinylthio acetic acid, acetic acid, 2-pyrimidinylthio, 2-carboxymethylthio pyrimidine, 2-pyrimidin-2-ylsulfanyl acetic acid, 2-pyrimidin-2-ylthioacetic acid, acetic acid, 2-2-pyrimidinylthio
581022
2-(pyrimidin-2-ylsulfanyl)acetate
[O-]C(=O)CSC1=NC=CC=N1
This item is not returnable. View return policy
88768-45-0
C6H5N2O2S
169.18
MFCD00010287
NIEOYUNNKKAQKI-UHFFFAOYSA-M
2-pyrimidylthio acetic acid, pyrimidin-2-ylthio acetic acid, 2-pyrimidin-2-ylthio acetic acid, pyrimidin-2-ylsulfanyl acetic acid, 2-pyrimidinylthio acetic acid, acetic acid, 2-pyrimidinylthio, 2-carboxymethylthio pyrimidine, 2-pyrimidin-2-ylsulfanyl acetic acid, 2-pyrimidin-2-ylthioacetic acid, acetic acid, 2-2-pyrimidinylthio
581022
2-(pyrimidin-2-ylsulfanyl)acetate
[O-]C(=O)CSC1=NC=CC=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∼200°C (decomposition)
Quantity 5 g
Beilstein 136468
Formula Weight 170.19
Percent Purity 98%
Chemical Name or Material 2- (Carboxymethylthio)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'