missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Carboxy-4-fluorobenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62176.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407126

  • £333.10 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Carboxy-4-fluorobenzeneboronic acid pinacol ester
266.07
2-Carboxy-4-fluorophenylboronic acid pinacol ester

Specifications

2-Carboxy-4-fluorobenzeneboronic acid pinacol ester
Solid
266.07
1g
96%
2-Carboxy-4-fluorophenylboronic acid pinacol ester
Videos
SDS