missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Carbomethoxy-3-thiophenesulfonyl chloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 62.0°C to 66.0°C |
Color | White to Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 1 g |
Formula Weight | 240.69 |
Percent Purity | 95% |
Show More |
Safety and Handling
Product Identifier
- 2-Carbomethoxy-3-thiophenesulfonyl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'