missing translation for 'onlineSavingsMsg'
Learn More

2-Carbomethoxy-3-thiophenesulfonyl chloride, 95%, Thermo Scientific™

Catalog Number 10726115
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
59337-92-7
C6H5ClO4S2
240.67
MFCD00068160
PJVJBDAUWILEOG-UHFFFAOYSA-N
methyl 3-chlorosulfonyl thiophene-2-carboxylate, 2-carbomethoxy-3-thiophenesulfonyl chloride, methylchlorosulfonylthiophenecarboxylate, methyl 3-chlorosulfonyl-2-thiophenecarboxylate, 3-chlorosulfonyl-2-thiophene carboxylic acid methyl ester, methyl-3-chlorosulfonylthiophene-2-carboxylate, 2-methoxycarbonyl thiophene-3-sulfonyl chloride, 2-thiophenecarboxylic acid, 3-chlorosulfonyl-, methyl ester, 2-methoxycarbonyl-3-thiophenesulfonyl chloride, 2-methoxycarbonyl thiophene-3-sulphonyl chloride
2736733
methyl 3-chlorosulfonylthiophene-2-carboxylate
COC(=O)C1=C(C=CS1)S(Cl)(=O)=O
This item is not returnable. View return policy
59337-92-7
C6H5ClO4S2
240.67
MFCD00068160
PJVJBDAUWILEOG-UHFFFAOYSA-N
methyl 3-chlorosulfonyl thiophene-2-carboxylate, 2-carbomethoxy-3-thiophenesulfonyl chloride, methylchlorosulfonylthiophenecarboxylate, methyl 3-chlorosulfonyl-2-thiophenecarboxylate, 3-chlorosulfonyl-2-thiophene carboxylic acid methyl ester, methyl-3-chlorosulfonylthiophene-2-carboxylate, 2-methoxycarbonyl thiophene-3-sulfonyl chloride, 2-thiophenecarboxylic acid, 3-chlorosulfonyl-, methyl ester, 2-methoxycarbonyl-3-thiophenesulfonyl chloride, 2-methoxycarbonyl thiophene-3-sulphonyl chloride
2736733
methyl 3-chlorosulfonylthiophene-2-carboxylate
COC(=O)C1=C(C=CS1)S(Cl)(=O)=O
CAS Min % 94.0
CAS Max % 100.0
Melting Point 62.0°C to 66.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 240.69
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Carbomethoxy-3-thiophenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Carbomethoxy-3-thiophenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'