missing translation for 'onlineSavingsMsg'
Learn More

2-Butyn-1-ol, 98%, Thermo Scientific Chemicals

Catalog Number p-80766
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
764-01-2
C4H6O
70.09
MFCD00002914
NEEDEQSZOUAJMU-UHFFFAOYSA-N
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
12991
but-2-yn-1-ol
CC#CCO
This item is not returnable. View return policy
764-01-2
C4H6O
70.09
MFCD00002914
NEEDEQSZOUAJMU-UHFFFAOYSA-N
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
12991
but-2-yn-1-ol
CC#CCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -2.2°C
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 142.0°C to 143.0°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C≡CCH2OH
Refractive Index 1.4520 to 1.4540
Quantity 5 mL
Beilstein 01, II, 505
Specific Gravity 0.93
Formula Weight 70.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Butyn-1-ol, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Butyn-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'