missing translation for 'onlineSavingsMsg'
Learn More

(2-Butyl-5-chloro-1H-imidazol-4-yl)methanol, 97%, Thermo Scientific™

Catalog Number p-2695058
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
79047-41-9
C8H13ClN2O
188.655
MFCD01934397
DXSZKDOOHOBZMT-UHFFFAOYSA-N
2-butyl-4-chloro-1h-imidazol-5-yl methanol, 2-butyl-5-chloro-1h-imidazol-4-yl methanol, 2-butyl-5-chloroimidazole-4-methanol, 2-butyl-4-chloro-5-hydroxymethyl imidazole, 2-butyl-4-chloro-1h-imidazole-5-methanol, 2-butyl-5-chloro-1h-imidazol-4-yl-methanol, 2-butyl-5-chloro-3h-imidazol-4-yl methanol, 2-butyl-4-chloro-5-hydroxyimidazole, losartan impurity 1, acmc-1bk6v
2735662
(2-butyl-4-chloro-1H-imidazol-5-yl)methanol
CCCCC1=NC(=C(N1)CO)Cl
This item is not returnable. View return policy
79047-41-9
C8H13ClN2O
188.655
MFCD01934397
DXSZKDOOHOBZMT-UHFFFAOYSA-N
2-butyl-4-chloro-1h-imidazol-5-yl methanol, 2-butyl-5-chloro-1h-imidazol-4-yl methanol, 2-butyl-5-chloroimidazole-4-methanol, 2-butyl-4-chloro-5-hydroxymethyl imidazole, 2-butyl-4-chloro-1h-imidazole-5-methanol, 2-butyl-5-chloro-1h-imidazol-4-yl-methanol, 2-butyl-5-chloro-3h-imidazol-4-yl methanol, 2-butyl-4-chloro-5-hydroxyimidazole, losartan impurity 1, acmc-1bk6v
2735662
(2-butyl-4-chloro-1H-imidazol-5-yl)methanol
CCCCC1=NC(=C(N1)CO)Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 188.66
Percent Purity 97%
Chemical Name or Material (2-Butyl-5-chloro-1H-imidazol-4-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'