Alfa Aesar™ 2-Buten-1-ol, cis + trans (ca 1:19), 96%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: A10681.14

2-BUTEN-1-OL 25G

UNSPSC: 12352104

Code: NEW

Additional Details:
Additional Details: CAS Number: 6117-91-5 Weight: 0.01000kg


Quantity 25g

Product Code. 11453444

Quantity Price
1 £ 47.59 / 25g
Estimated Shipment date
from Supplier 09-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Buten-1-ol
CAS 6117-91-5
Name Note cis + trans (≈ 1:19)
Molecular Formula CH3CH=CHCH2OH
Formula Weight 72.11
MDL Number MFCD00002923
UN Number UN1987
Beilstein 1719374
Merck Index 14,2601
Synonym Crotyl alcohol; 1-Hydroxybut-2-ene

Specification

Formula Weight 72.11
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Buten-1-ol
TSCA TSCA
Name Note cis + trans (≈ 1:19)
Assay 96%
Molecular Formula CH3CH=CHCH2OH
MDL Number MFCD00002923
CAS 6117-91-5
UN Number UN1987
Beilstein 1719374
Merck Index 14,2601
Synonym Crotyl alcohol; 1-Hydroxybut-2-ene
Boiling Point 121° to 122°C
Quantity 25g
Flash Point 37°C (98°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H302-H312
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.
RTECS Number EM9275000

Applications
2-Buten-1-ol, cis + trans (ca 1:19) is used as a building block for the synthesis of various pharmaceutical compounds such as crotyl mercaptan.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, strong oxidizing agents, strong reducing agents, acid chlorides, acid anhydrides and phosphorus halides.

Safety and Handling

Warning

GHS H Statement
H226-H302-H312
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.