missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Buten-1-ol, cis + trans (ca 1:19), 96%

Brand:  Alfa Aesar™ A10681.22

Code : NEW

Additional Details : CAS Number : 6117-91-5 Weight : 0.10000kg Transport : UN number : 1987 Chem class : 3 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 11463444

  • £143.10 / 100g


Estimated Shipment
14-06-2018
Add to basket

Description

Description

Applications
2-Buten-1-ol, cis + trans (ca 1:19) is used as a building block for the synthesis of various pharmaceutical compounds such as crotyl mercaptan.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, strong oxidizing agents, strong reducing agents, acid chlorides, acid anhydrides and phosphorus halides.
Specifications

Chemical Identifiers

2-Buten-1-ol
cis + trans (≈ 1:19)
72.11
UN1987
14,2601
6117-91-5
CH3CH=CHCH2OH
MFCD00002923
1719374
Crotyl alcohol; 1-Hydroxybut-2-ene

Specifications

2-Buten-1-ol
6117-91-5
CH3CH=CHCH2OH
MFCD00002923
1719374
Crotyl alcohol; 1-Hydroxybut-2-ene
100g
cis + trans (≈ 1:19)
96%
72.11
UN1987
14,2601
121° to 122°C
37°C (98°F)
Videos
SDS