missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, 99+%, for electronic use, residue free, Thermo Scientific Chemicals

Catalog Number p-3737777
28.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
78-93-3
C4H8O
72.11
MFCD00011648
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
6569
CHEBI:28398
butan-2-one
CCC(C)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
78-93-3
C4H8O
72.11
MFCD00011648
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
6569
CHEBI:28398
butan-2-one
CCC(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -87.0°C
CAS Min % 99.0
CAS Max % 100.0
Density 0.8060g/mL
Boiling Point 80.0°C
Quantity 1 L
Flash Point −7°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3780 to 1.3800
Assay Percent Range 99+%
Residue after Evaporation 0.0005% max.
Specific Gravity 0.806
Linear Formula C2H5COCH3
Viscosity 0.42 mPa.s (15°C)
Beilstein 01, IV, 3243
Water 0.2% max. (Coulometric)
Fieser 01,678
Merck Index 15, 6143
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: soluble in benzene, alcohol and ether, miscible with oils
Formula Weight 72.11
Percent Purity ≥99%
Grade Electronic
Chemical Name or Material 2-Butanone, Residue Free
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'