missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone, 97%, Thermo Scientific Chemicals

Catalog Number p-7049906
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
This item is not returnable. View return policy
78-93-3
C4H8O
72.11
MFCD00011648
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
6569
CHEBI:28398
butan-2-one
CCC(C)=O
This item is not returnable. View return policy
78-93-3
C4H8O
72.11
MFCD00011648
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
6569
CHEBI:28398
butan-2-one
CCC(C)=O

2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -87°C
Color Colorless
Density 0.805 g/mL
Boiling Point 80°C
Quantity 1 L
Flash Point -3°C (26°F)
Odor Characteristic
Physical Form Liquid
Refractive Index 1.379
Linear Formula CH3COCH2CH3
UN Number UN1193
Beilstein 741880
Merck Index 14,6072
Solubility Information Miscible with cold water,diethyl ether and acetone.
Formula Weight 72.11
Percent Purity 97%
Chemical Name or Material 2-Butanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'