missing translation for 'onlineSavingsMsg'
Learn More

2-Bromotoluene, 99%, Thermo Scientific Chemicals

Catalog Number p-77256
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
95-46-5
C7H7Br
171.04
MFCD00000068
QSSXJPIWXQTSIX-UHFFFAOYSA-N
2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide
7236
1-bromo-2-methylbenzene
CC1=CC=CC=C1Br
This item is not returnable. View return policy
95-46-5
C7H7Br
171.04
MFCD00000068
QSSXJPIWXQTSIX-UHFFFAOYSA-N
2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide
7236
1-bromo-2-methylbenzene
CC1=CC=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -27°C
Color Colorless to Yellow
Density 1.4200g/mL
Boiling Point 181.5°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4Br
Refractive Index 1.5550 to 1.557
Quantity 25 mL
Beilstein 05, 304
Merck Index 14, 1439
Specific Gravity 1.42
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with alcohol and benzene
Formula Weight 171.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromotoluene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'