missing translation for 'onlineSavingsMsg'
Learn More

2-Bromotoluene, 99%, Thermo Scientific Chemicals

Catalog Number p-7082064
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
95-46-5
C7H7Br
171.04
MFCD00000068
QSSXJPIWXQTSIX-UHFFFAOYSA-N
2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide
7236
1-bromo-2-methylbenzene
CC1=CC=CC=C1Br
This item is not returnable. View return policy
95-46-5
C7H7Br
171.04
MFCD00000068
QSSXJPIWXQTSIX-UHFFFAOYSA-N
2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide
7236
1-bromo-2-methylbenzene
CC1=CC=CC=C1Br

2-Bromotoluene was used in the synthesis of (±)-isocomene. Suzuki coupling of 2-bromotoluene with phenylboronic acid using palladium nanoparticles supported on alumina-based oxides as catalyst has been reported. 2-Bromotoluene is used in manufacturing pharmaceuticals (especially for nonsteroidal antiinflammatory drugs; Meclofenamic acid, Mefenamic acid) and other organic compounds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromotoluene was used in the synthesis of (±)-isocomene. Suzuki coupling of 2-bromotoluene with phenylboronic acid using palladium nanoparticles supported on alumina-based oxides as catalyst has been reported. 2-Bromotoluen
TRUSTED_SUSTAINABILITY
Melting Point -27°C
Density 1.42
Boiling Point 179°C to 181°C
Flash Point 78°C (172°F)
Refractive Index 1.556
Quantity 25 g
Beilstein 1904176
Merck Index 14,1439
Solubility Information Soluble in alcohol,ether,acetone,and ethanol. Insoluble in water.
Formula Weight 171.04
Percent Purity 99%
Chemical Name or Material 2-Bromotoluene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'