missing translation for 'onlineSavingsMsg'
Learn More

2-Bromothiazole, 98+%, Thermo Scientific Chemicals

Catalog Number p-77228
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
3034-53-5
C3H2BrNS
164.03
MFCD00005316
RXNZFHIEDZEUQM-UHFFFAOYSA-N
2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
76430
2-bromo-1,3-thiazole
C1=CSC(=N1)Br
This item is not returnable. View return policy
3034-53-5
C3H2BrNS
164.03
MFCD00005316
RXNZFHIEDZEUQM-UHFFFAOYSA-N
2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol
76430
2-bromo-1,3-thiazole
C1=CSC(=N1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Brown-Orange to Colorless
Density 1.8200g/mL
Boiling Point 70°C to 71°C (20.0 mmHg)
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5910 to 1.593
Quantity 5 mL
Beilstein 27, 15
Specific Gravity 1.82
Solubility Information Solubility in water: insoluble
Formula Weight 164.03
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Bromothiazole, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromothiazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'