missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoterephthalic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-2689963
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
586-35-6
C8H5BrO4
245.03
MFCD00002403
QPBGNSFASPVGTP-UHFFFAOYSA-N
bromoterephthalic acid, 2-bromobenzene-1,4-dicarboxylic acid, 2-bromoterephthalicacid, 2-bromo-terephthalic acid, 2-bromo-1,4-benzenedicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-bromo, labotest-bb lt00454996, pubchem12798, 2-bromo terephthalic acid, acmc-1aoy9
68513
2-bromoterephthalic acid
C1=CC(=C(C=C1C(=O)O)Br)C(=O)O
This item is not returnable. View return policy
586-35-6
C8H5BrO4
245.03
MFCD00002403
QPBGNSFASPVGTP-UHFFFAOYSA-N
bromoterephthalic acid, 2-bromobenzene-1,4-dicarboxylic acid, 2-bromoterephthalicacid, 2-bromo-terephthalic acid, 2-bromo-1,4-benzenedicarboxylic acid, 1,4-benzenedicarboxylic acid, 2-bromo, labotest-bb lt00454996, pubchem12798, 2-bromo terephthalic acid, acmc-1aoy9
68513
2-bromoterephthalic acid
C1=CC(=C(C=C1C(=O)O)Br)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 295.0°C to 297.0°C
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula BrC6H3(CO2H)2
Quantity 25 g
Beilstein 09,848
Formula Weight 245.03
Percent Purity 95%
Chemical Name or Material 2-Bromoterephthalic acid, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromoterephthalic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'