missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromopyridine-5-carboxaldehyde, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.
Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. 2-7°C Refrigerate.
A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.
Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. 2-7°C Refrigerate.
Specifications
Melting Point | 104°C to 110°C (decomposition) |
Color | Yellow |
Quantity | 1 g |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in dichloromethane,ether,ethyl acetate and methanol. |
Formula Weight | 186.01 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 2-Bromopyridine-5-carboxaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'