missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopyridine-4-boronic acid, 95%, Thermo Scientific Chemicals

Catalog Number 11966361
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
458532-94-0
C5H5BBrNO2
201.81
MFCD06798244
DFIKFQMGZNTFLI-UHFFFAOYSA-N
2-bromopyridine-4-boronic acid, 2-bromopyridin-4-yl boronic acid, boronic acid, 2-bromo-4-pyridinyl-9ci, 2-bromo-4-pyridyl boronic acid, 2-bromo-4-pyridineboronic acid, boronicacid, 2-bromo-4-pyridinyl-9ci, acmc-209k4o, ksc234s1f, 2-bromo-4-pyridinyl boronic acid, 2-bromanylpyridin-4-yl boronic acid
12060162
(2-bromopyridin-4-yl)boronic acid
OB(O)C1=CC(Br)=NC=C1
This item is not returnable. View return policy
458532-94-0
C5H5BBrNO2
201.81
MFCD06798244
DFIKFQMGZNTFLI-UHFFFAOYSA-N
2-bromopyridine-4-boronic acid, 2-bromopyridin-4-yl boronic acid, boronic acid, 2-bromo-4-pyridinyl-9ci, 2-bromo-4-pyridyl boronic acid, 2-bromo-4-pyridineboronic acid, boronicacid, 2-bromo-4-pyridinyl-9ci, acmc-209k4o, ksc234s1f, 2-bromo-4-pyridinyl boronic acid, 2-bromanylpyridin-4-yl boronic acid
12060162
(2-bromopyridin-4-yl)boronic acid
OB(O)C1=CC(Br)=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 201.82
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromopyridine-4-boronic acid
compliance-icons
Product Identifier
  • 2-Bromopyridine-4-boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'