missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromopropionyl bromide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
2-Bromopropionyl bromide is used in the preparation of cellulose macroinitiator through direct acylation of cellulose in ionic liquid 1-allyl-3-methylimidazolium chloride. It is also used to prepare 3-(2-bromopropionyl)-2-oxazolidone derivatives and bromoester terminated poly(3-hexylthiophene). Further, it is used as a macroinitiator for atom transfer radical polymerization of acrylates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromopropionyl bromide is used in the preparation of cellulose macroinitiator through direct acylation of cellulose in ionic liquid 1-allyl-3-methylimidazolium chloride. It is also used to prepare 3-(2-bromopropionyl)-2-oxazolidone derivatives and bromoester terminated poly(3-hexylthiophene). Further, it is used as a macroinitiator for atom transfer radical polymerization of acrylates.
Solubility
Miscible with trichloromethane, diethyl ether, benzene and acetone.
Notes
Incompatible with water, alcohols, oxidizing agents and strong alkalis.
2-Bromopropionyl bromide is used in the preparation of cellulose macroinitiator through direct acylation of cellulose in ionic liquid 1-allyl-3-methylimidazolium chloride. It is also used to prepare 3-(2-bromopropionyl)-2-oxazolidone derivatives and bromoester terminated poly(3-hexylthiophene). Further, it is used as a macroinitiator for atom transfer radical polymerization of acrylates.
Solubility
Miscible with trichloromethane, diethyl ether, benzene and acetone.
Notes
Incompatible with water, alcohols, oxidizing agents and strong alkalis.
Specifications
Density | 2.061 |
Boiling Point | 48°C to 50°C (10 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.521 |
Quantity | 100 g |
UN Number | UN3265 |
Beilstein | 1071331 |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible with trichloromethane,diethyl ether,benzene and acetone. |
Formula Weight | 215.88 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'