missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number 10778663
126.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
244205-40-1
C6H6BBrO2
200.83
MFCD01114672
PLVCYMZAEQRYHJ-UHFFFAOYSA-N
2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid
2773294
(2-bromophenyl)boronic acid
OB(O)C1=CC=CC=C1Br
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
244205-40-1
C6H6BBrO2
200.83
MFCD01114672
PLVCYMZAEQRYHJ-UHFFFAOYSA-N
2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid
2773294
(2-bromophenyl)boronic acid
OB(O)C1=CC=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 109.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula BrC6H4B(OH)2
Quantity 25 g
Formula Weight 200.83
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Bromophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'