missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromophenyl acetate, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Assay Percent Range | 97.5 % min. (GC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 215.05 |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromophenyl acetate |
Safety and Handling
Product Identifier
- 2-Bromophenyl acetate, 98%
- Warning
- Acute toxicity Category 4
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'