missing translation for 'onlineSavingsMsg'
Learn More

2-Bromopentane, 90%, tech., Thermo Scientific Chemicals

Catalog Number 10766321
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
107-81-3
C5H11Br
151.05
MFCD00000160
LGAJYTCRJPCZRJ-UHFFFAOYSA-N
pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j
7890
2-bromopentane
CCCC(C)Br
This item is not returnable. View return policy
107-81-3
C5H11Br
151.05
MFCD00000160
LGAJYTCRJPCZRJ-UHFFFAOYSA-N
pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j
7890
2-bromopentane
CCCC(C)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Melting Point -95.5°C
Density 1.2200g/mL
Boiling Point 116.0°C to 117.0°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 12% max. (3-Bromopentane) Typical (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2CH(Br)CH3
Refractive Index 1.4393 to 1.4413
Quantity 100 mL
Beilstein 01, 131
Specific Gravity 1.22
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with organic solvents
Formula Weight 151.05
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material 2-Bromopentane, tech., 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromopentane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'