missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromonitrobenzene-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Bromo-2-nitrobenze-d4 |
| CAS | 1020720-09-5 |
| Molecular Formula | C6 2H4 Br N O2 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C6H4BrNO2/c7-5-3-1-2-4-6(5)8(9)10/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c(Br)c1[2 H])[N+](=O)[O-] |
| IUPAC Name | 1-bromo-2,3,4,5-tetradeuterio-6-nitrobenzene |
| Molecular Weight (g/mol) | 206.03 |
| Formula Weight | 204.9676 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction