missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromoisophthalaldehyde, 97%

Brand:  Alfa Aesar™ H64423.MD

Code : NEW

Additional Details : CAS Number : 79839-49-9 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15431629

  • £13.60 / 250mg


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Description

Description

Applications
Used in the preparation of paclitaxel-mimicking alkaloids with anticancer activity.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2-Bromoisophthalaldehyde
213.03
4740331
79839-49-9
MFCD08461034
2-Bromobenzene-1,3-dicarboxaldehyde

Specifications

2-Bromoisophthalaldehyde
139° to 144°C
Solid
213.03
4740331
79839-49-9
250mg
97%
MFCD08461034
2-Bromobenzene-1,3-dicarboxaldehyde
Videos
SDS