missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoisonicotinic acid, 97%, Thermo Scientific™

Catalog Number 10882261
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
66572-56-3
C6H4BrNO2
202.01
YBTKGKVQEXAYEM-UHFFFAOYSA-N
2-bromoisonicotinic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromo-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 2-bromo, 2-bromoisonicotic acid, 2-bromisonicotins, 2-bromopyridine-4-carboxylicacid, pubchem9211, acmc-209nvt
222701
2-bromopyridine-4-carboxylic acid
C1=CN=C(C=C1C(=O)O)Br
This item is not returnable. View return policy
66572-56-3
C6H4BrNO2
202.01
YBTKGKVQEXAYEM-UHFFFAOYSA-N
2-bromoisonicotinic acid, 2-bromo-4-pyridinecarboxylic acid, 2-bromo-isonicotinic acid, 2-bromo-4-pyridine carboxylic acid, 4-pyridinecarboxylic acid, 2-bromo, 2-bromoisonicotic acid, 2-bromisonicotins, 2-bromopyridine-4-carboxylicacid, pubchem9211, acmc-209nvt
222701
2-bromopyridine-4-carboxylic acid
C1=CN=C(C=C1C(=O)O)Br
CAS Min % 96.0
CAS Max % 100.0
Melting Point 212.0°C to 222.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 202.01
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Bromoisonicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromoisonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'