missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromofuran, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5, 0.05, 0.45, 0.35 |
CAS Max % | 98.5, 0.15, 0.55, 0.45 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 146.97 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromofuran, stabilized with Copper (0.1%), Furan (0.5%) and Sodium Bicarbonate (0.4%) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'