missing translation for 'onlineSavingsMsg'
Learn More

2-Bromofluorene, 95%, Thermo Scientific Chemicals

Catalog Number p-3735739
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
1133-80-8
MFCD00001115
FXSCJZNMWILAJO-UHFFFAOYSA-N
2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl
14336
2-bromo-9H-fluorene
C1C2=CC=CC=C2C3=C1C=C(C=C3)Br
This item is not returnable. View return policy
1133-80-8
MFCD00001115
FXSCJZNMWILAJO-UHFFFAOYSA-N
2-bromofluorene, 9h-fluorene, 2-bromo, 2-brom-9h-fluoren, 2-bromofluorene, tech., #, acmc-1c7b2, ksc181a7p, 2-bromofluorene 5g, 4-bromo-2,2'-methylenebiphenyl
14336
2-bromo-9H-fluorene
C1C2=CC=CC=C2C3=C1C=C(C=C3)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 110°C to 115°C
Color White to Yellow
Boiling Point 185°C (135.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C13H9Br
Quantity 25 g
Beilstein 05, II, 534
Molecular Weight (g/mol) 245.12
Formula Weight 245.12
Percent Purity 95%
Physical Form Slightly Crystalline Powder or Flakes
Chemical Name or Material 2-Bromofluorene
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromofluorene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'