missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethylamine Hydrobromide 99%, ACROS Organics™

Brand:  Acros Organics 106930025

Code : 29

Additional Details : CAS Number : 2576-47-8 Weight : 2.55000kg

Quantity 2.5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 11905171

  • £379.40 / 2.50kg


EU Stock 3
Add to basket

Specifications

Chemical Identifiers

2-Bromoethylamine hydrobromide
107-09-5
100.0
99%
BrCH2CH2NH2·HBr
MFCD00012886
99%
98.5
99%
C2H6BrN·HBr
204.9

Specifications

2-Bromoethylamine hydrobromide
107-09-5
100.0
Plastic bottle
White to light beige
Crystalline Powder or Crystals
99%
C2H6BrN·HBr
204.9
99%
98.5
Authentic
this product has a strong avidity for water
172.0° to 176.0°C
99%
2.5kg
BrCH2CH2NH2·HBr
MFCD00012886
Videos
SDS