missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl methyl ether, 95%, ACROS Organics™

Brand:  Acros Organics 402981000

Code : NEW

Additional Details : CAS Number : 6482-24-2 Weight : 0.10000kg Transport : UN number : 1993 Chem class : 3 Pack group : III


Packaging Glass bottle
Quantity 100g

 View more versions of this product

Product Code. 13458548

  • £147.40 / 100g

EU Stock 2

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

6482-24-2
138.99
YZUPZGFPHUVJKC-UHFFFAOYSA-N
80972
COCCBr
C3H7BrO
MFCD00000236
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane
1-bromo-2-methoxyethane

Specifications

2-Bromoethyl methyl ether
28°C
Glass bottle
1.46
Colorless to Yellow
6482-24-2
100.0
C3H7BrO
MFCD00000236
Solubility in water: 14.4g/L (25°C)
COCCBr
138.99
138.99
95%
1.4600g/mL
Authentic
1.4460 to 1.4500
107.0°C to 110.0°C
100g
94.0
94% min. (GC)
BrCH2CH2OCH3
2-bromoethyl methyl ether, 2-methoxyethyl bromide, ethane, 1-bromo-2-methoxy, ether, 2-bromoethyl methyl, methoxyethyl bromide, 2-bromoethylmethylether, bromoethyl methyl ether, 2-bromoethylmethyl ether, 2-bromoethylmethyl-ether, 2-bromo-1-methoxyethane
YZUPZGFPHUVJKC-UHFFFAOYSA-N
1-bromo-2-methoxyethane
80972
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bromoethyl methyl ether

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documents