missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethanol, 97%, Thermo Scientific Chemicals

Catalog Number 10616292
Click to view available options
Quantity:
5 mL
100 mL
250 mL
1 L
Packaging:
Glass bottle
540-51-2
C2H5BrO
124.96
MFCD00002827
LDLCZOVUSADOIV-UHFFFAOYSA-N
ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol
10898
2-bromoethanol
C(CBr)O
540-51-2
C2H5BrO
124.96
MFCD00002827
LDLCZOVUSADOIV-UHFFFAOYSA-N
ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol
10898
2-bromoethanol
C(CBr)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -80.0°C
Color Colorless to Yellow
Density 1.7600g/mL
Boiling Point 149.0°C to 150.0°C (750.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CH2OH
Refractive Index 1.4900 to 1.4940
Quantity 250 mL
Beilstein 01, 338
Fieser 11,77
Merck Index 15, 3846
Specific Gravity 1.76
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with ethanol, ether, soluble in most common organic solvents,, except petroleum ether
Formula Weight 124.96
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromoethanol, 97%
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'