missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobutyric acid, 99%, Thermo Scientific Chemicals

Catalog Number 10513341
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
80-58-0
C4H7BrO2
167.00
MFCD00004216,MFCD00210111,MFCD00210112
YAQLSKVCTLCIIE-UHFFFAOYNA-N
2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid
6655
2-bromobutanoic acid
CCC(Br)C(O)=O
This item is not returnable. View return policy
80-58-0
C4H7BrO2
167.00
MFCD00004216,MFCD00210111,MFCD00210112
YAQLSKVCTLCIIE-UHFFFAOYNA-N
2-bromobutyric acid, alpha-bromobutyric acid, butanoic acid, 2-bromo, butyric acid, 2-bromo, .alpha.-bromobytyric acid, butyric acid, .alpha.-bromo, dl-2-bromobutyric acid, a-bromobutyric acid, butyric acid, alpha-bromo, bromobutyric acid
6655
2-bromobutanoic acid
CCC(Br)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula C2H5CH(Br)CO2H
Refractive Index 1.4712 to 1.4732 (20°C, 589nm)
Quantity 250 mL
Beilstein 02, 281
Merck Index 15, 1420
Formula Weight 167
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 2-Bromobutyric acid, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromobutyric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'