missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobornane, technical, Thermo Scientific Chemicals

Catalog Number p-7077853
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4443-48-5
C10H17Br
217.15
OTOQMOVZIUGCQE-UHFFFAOYSA-N
bornyl bromide, 2-bromobornane, bornyl bromide, pract., 2-bromo-1,7,7-trimethylbicyclo 2.2.1 heptane, bornylbromid, bornylbromide, bornane, 2-bromo, 3-bromo-4,7,7-trimethylbicyclo 2.2.1 heptane, 2-bromo-1,7,7-trimethylbicyclo 2.2.1 heptane #
138249
3-bromo-4,7,7-trimethylbicyclo[2.2.1]heptane
CC1(C2CCC1(C(C2)Br)C)C
This item is not returnable. View return policy
4443-48-5
C10H17Br
217.15
OTOQMOVZIUGCQE-UHFFFAOYSA-N
bornyl bromide, 2-bromobornane, bornyl bromide, pract., 2-bromo-1,7,7-trimethylbicyclo 2.2.1 heptane, bornylbromid, bornylbromide, bornane, 2-bromo, 3-bromo-4,7,7-trimethylbicyclo 2.2.1 heptane, 2-bromo-1,7,7-trimethylbicyclo 2.2.1 heptane #
138249
3-bromo-4,7,7-trimethylbicyclo[2.2.1]heptane
CC1(C2CCC1(C(C2)Br)C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 94.0°C to 96.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 217.15
Percent Purity ≥78.0% (GC)
Grade Technical
Chemical Name or Material 2-Bromobornane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'