missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzylamine, 98%, Thermo Scientific Chemicals

Catalog Number 10282892
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3959-05-5
C7H8BrN
186.05
MFCD00025572
NOYASZMZIBFFNZ-UHFFFAOYSA-N
2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo
334072
(2-bromophenyl)methanamine
C1=CC=C(C(=C1)CN)Br
This item is not returnable. View return policy
3959-05-5
C7H8BrN
186.05
MFCD00025572
NOYASZMZIBFFNZ-UHFFFAOYSA-N
2-bromobenzylamine, 2-bromophenyl methanamine, o-bromobenzylamine, 2-bromophenyl methylamine, 1-2-bromophenyl methanamine, 2-brobenzylamine, 2-bromo-benzylamine, pubchem20398, acmc-1ct06, benzenemethanamine, 2-bromo
334072
(2-bromophenyl)methanamine
C1=CC=C(C(=C1)CN)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH2NH2
Refractive Index 1.5875 to 1.5910
Quantity 1 g
Beilstein 12,III,2348
Formula Weight 186.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromobenzylamine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromobenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'