Learn More
2-Bromobenzotrifluoride, 99%, ACROS Organics™
Brand: Acros Organics 106731000
Code : 29
Additional Details : CAS Number : 392-83-6 Weight : 0.15000kg Transport : UN number : 1993 Chem class : 1 Pack group : III
Quantity | 100g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
2-Bromobenzotrifluoride | |
98.5 | |
≥98.5% | |
C7H4BrF3 | |
225.01 | |
o-Bromobenzotrifluoride |
392-83-6 | |
100.0 | |
99% | |
BrC6H4CF3 | |
MFCD00000373 |
Specifications
2-Bromobenzotrifluoride | |
Authentic | |
100.0 | |
1.4810 to 1.4830 (20°C, 589nm) | |
100g | |
≥98.5% | |
C7H4BrF3 | |
225.01 | |
o-Bromobenzotrifluoride |
392-83-6 | |
98.5 | |
Glass bottle | |
Clear colorless to brown | |
Liquid | |
99% | |
BrC6H4CF3 | |
MFCD00000373 |