missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzaldehyde, 97%, ACROS Organics™

Brand:  Acros Organics 106650250

Code : 29

Additional Details : CAS Number : 6630-33-7 Weight : 0.04000kg


Quantity 25mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10479200

  • £26.60 / 25mL


EU Stock 4
Add to basket


Specifications

Chemical Identifiers

2-Bromobenzaldehyde
96.0
97%
C7H5BrO
185.02
6630-33-7
100.0
97%
BrC6H4CHO

Specifications

2-Bromobenzaldehyde
Authentic
100.0
1.5950 to 1.5970
Clear yellow
230.0°C
95°C
97%
C7H5BrO
185.02
6630-33-7
96.0
Glass bottle
1.5800g/mL
21.0° to 22.0°C
25mL
Liquid After Melting
97%
BrC6H4CHO
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bromobenzaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS