missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-4229101
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
6630-33-7
C7H5BrO
185.02
MFCD00003300
NDOPHXWIAZIXPR-UHFFFAOYSA-N
o-bromobenzaldehyde, benzaldehyde, 2-bromo, benzaldehyde, o-bromo, 2-bromo benzaldehyde, bromobenzaldehyde, bromo benzaldehyde, o-bromo-benzaldehyde, 2-formylbromobenzene, 2-bromo-benzaldehyde, benzaldehyde, bromo
81129
2-bromobenzaldehyde
BrC1=CC=CC=C1C=O
This item is not returnable. View return policy
6630-33-7
C7H5BrO
185.02
MFCD00003300
NDOPHXWIAZIXPR-UHFFFAOYSA-N
o-bromobenzaldehyde, benzaldehyde, 2-bromo, benzaldehyde, o-bromo, 2-bromo benzaldehyde, bromobenzaldehyde, bromo benzaldehyde, o-bromo-benzaldehyde, 2-formylbromobenzene, 2-bromo-benzaldehyde, benzaldehyde, bromo
81129
2-bromobenzaldehyde
BrC1=CC=CC=C1C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 21.0°C to 22.0°C
Color Yellow
Density 1.5800g/mL
Boiling Point 230.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CHO
Refractive Index 1.5950 to 1.5970
Quantity 10 mL
Beilstein 07,238
Specific Gravity 1.58
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 185.02
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 2-Bromobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'