missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoanisole, 97%, Thermo Scientific Chemicals

Catalog Number p-71551
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
578-57-4
C7H7BrO
187.04
MFCD00000064
HTDQSWDEWGSAMN-UHFFFAOYSA-N
2-bromoanisole, o-bromoanisole, benzene, 1-bromo-2-methoxy, 2-methoxybromobenzene, o-anisyl bromide, anisole, o-bromo, o-methoxybromobenzene, 2-bromoanisol, o-methoxyphenyl bromide, 2-methoxyphenyl bromide
11358
1-bromo-2-methoxybenzene
COC1=CC=CC=C1Br
This item is not returnable. View return policy
578-57-4
C7H7BrO
187.04
MFCD00000064
HTDQSWDEWGSAMN-UHFFFAOYSA-N
2-bromoanisole, o-bromoanisole, benzene, 1-bromo-2-methoxy, 2-methoxybromobenzene, o-anisyl bromide, anisole, o-bromo, o-methoxybromobenzene, 2-bromoanisol, o-methoxyphenyl bromide, 2-methoxyphenyl bromide
11358
1-bromo-2-methoxybenzene
COC1=CC=CC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 2°C
Color Colorless to Yellow
Density 1.5000g/mL
Boiling Point 223°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4OCH3
Refractive Index 1.5730 to 1.575
Quantity 25 mL
Beilstein 06, 197
Specific Gravity 1.5
Solubility Information Solubility in water: immiscible
Formula Weight 187.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromoanisole, 97%
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'