missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromoaniline, 98%, Thermo Scientific Chemicals
19.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 29.0°C to 31.0°C |
Density | 1.5780g/mL |
Boiling Point | 229.0°C |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | BrC6H4NH2 |
Show More |
Safety and Handling
Product Identifier
- 2-Bromoaniline
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'