missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetophenone, 98%, Thermo Scientific Chemicals

Catalog Number 10669853
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
70-11-1
C8H7BrO
199.05
MFCD00000195
LIGACIXOYTUXAW-UHFFFAOYSA-N
2-bromoacetophenone, phenacyl bromide, 2-bromo-1-phenylethan-1-one, bromoacetophenone, ethanone, 2-bromo-1-phenyl, bromomethyl phenyl ketone, alpha-bromoacetophenone, stauffer 4644, omega-bromoacetophenone, acetophenone, 2-bromo
6259
CHEBI:51846
2-bromo-1-phenylethanone
C1=CC=C(C=C1)C(=O)CBr
This item is not returnable. View return policy
70-11-1
C8H7BrO
199.05
MFCD00000195
LIGACIXOYTUXAW-UHFFFAOYSA-N
2-bromoacetophenone, phenacyl bromide, 2-bromo-1-phenylethan-1-one, bromoacetophenone, ethanone, 2-bromo-1-phenyl, bromomethyl phenyl ketone, alpha-bromoacetophenone, stauffer 4644, omega-bromoacetophenone, acetophenone, 2-bromo
6259
CHEBI:51846
2-bromo-1-phenylethanone
C1=CC=C(C=C1)C(=O)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 46.0°C to 51.0°C
Color Brown-Green to White
Boiling Point 135.0°C (18.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5COCH2Br
Quantity 2.5 kg
Beilstein 07, 283
Fieser 02,316; 12,381
Merck Index 15, 1409
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alcohol,benzene,chloroform,and ether
Formula Weight 199.05
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 2-Bromoacetophenone, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromoacetophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'