missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-p-xylene, 97%, ACROS Organics™

Brand:  Acros Organics 173420250

Code : 29

Additional Details : CAS Number : 553-94-6 Weight : 0.07500kg


Quantity 25g

 View more versions of this product

Product Code. 10449470

  • £20.10 / 25g


EU Stock 8
Add to basket


Specifications

Chemical Identifiers

2-Bromo-p-xylene
553-94-6
100.0
97%
(CH3)2C6H3Br
MFCD00000074
97%
96.0
≥96.0%
C8H9Br
185.06
2, 5-Dimethylbromobenzene, 2-Bromo-1, 4-dimethylbenzene

Specifications

2-Bromo-p-xylene
553-94-6
100.0
Glass bottle
Liquid
97%
(CH3)2C6H3Br
MFCD00000074
Clear colorless to slightly colored
25g
97%
96.0
Authentic
1.5495 to 1.5515 (20°C, 589nm)
≥96.0%
C8H9Br
185.06
05, 385
2, 5-Dimethylbromobenzene, 2-Bromo-1, 4-dimethylbenzene
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bromo-p-xylene

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS