missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-N-methylbenzamide, 99%

£104.00 - £469.00

Chemical Identifiers

CAS 61436-88-2
Molecular Formula C8H8BrNO
Molecular Weight (g/mol) 214.062
MDL Number MFCD00099250
InChI Key FKVCJQNPXXXZIE-UHFFFAOYSA-N
Synonym n-methyl 2-bromobenzamide, n-methyl-2-bromobenzamide, benzamide,2-bromo-n-methyl, benzamide, 2-bromo-n-methyl, acmc-209msn, 2-bromo-n-methylbenzamide, 2-bromo-n-methyl-benzamide, 2-bromanyl-n-methyl-benzamide, 2-bromophenyl-n-methylcarboxamide
PubChem CID 264945
IUPAC Name 2-bromo-N-methylbenzamide
SMILES CNC(=O)C1=CC=CC=C1Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11430060 SDS View Documents Alfa Aesar™
H50583.03
1g £104.00
1g View alternative products
Save Up to 
»
EU Stock 2 Please to purchase this item. Need a web account? Register with us today!
11440060 SDS View Documents Alfa Aesar™
H50583.06
5g £469.00
5g View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

61436-88-2
214.062
FKVCJQNPXXXZIE-UHFFFAOYSA-N
264945
CNC(=O)C1=CC=CC=C1Br
C8H8BrNO
MFCD00099250
n-methyl 2-bromobenzamide, n-methyl-2-bromobenzamide, benzamide,2-bromo-n-methyl, benzamide, 2-bromo-n-methyl, acmc-209msn, 2-bromo-n-methylbenzamide, 2-bromo-n-methyl-benzamide, 2-bromanyl-n-methyl-benzamide, 2-bromophenyl-n-methylcarboxamide
2-bromo-N-methylbenzamide

Specifications

2-Bromo-N-methylbenzamide
61436-88-2
MFCD00099250
FKVCJQNPXXXZIE-UHFFFAOYSA-N
2-bromo-N-methylbenzamide
264945
99%
1g
C8H8BrNO
n-methyl 2-bromobenzamide, n-methyl-2-bromobenzamide, benzamide,2-bromo-n-methyl, benzamide, 2-bromo-n-methyl, acmc-209msn, 2-bromo-n-methylbenzamide, 2-bromo-n-methyl-benzamide, 2-bromanyl-n-methyl-benzamide, 2-bromophenyl-n-methylcarboxamide
CNC(=O)C1=CC=CC=C1Br
214.062
214.06