missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Bromo-N-(3-methylbenzyl)benzenesulfonamide, 97%
Brand: Alfa Aesar™ H60323.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
2-Bromo-N-(3-methylbenzyl)benzenesulfonamide | |
C14H14BrNO2S | |
340.23 | |
MFCD10634865 |
Specifications
2-Bromo-N-(3-methylbenzyl)benzenesulfonamide | |
C14H14BrNO2S | |
MFCD10634865 |
97% | |
340.23 | |
1g |