missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-N-(2,4-dichlorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60579.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15475856

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Bromo-N-(2,4-dichlorophenyl)benzenesulfonamide
C12H8BrCl2NO2S
381.07
MFCD12650431

Specifications

2-Bromo-N-(2,4-dichlorophenyl)benzenesulfonamide
C12H8BrCl2NO2S
MFCD12650431
97%
381.07
1g
Videos
SDS