missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-α-methylbenzyl alcohol 99%, ACROS Organics™

Brand:  Acros Organics 393640250

Code : 29

Additional Details : CAS Number : 5411-56-3 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10483105

  • £200.70 / 25g


Estimated Shipment
21-06-2018
Add to basket


Specifications

Chemical Identifiers

2-Bromo-α-methylbenzyl alcohol
98.5
≥98.5%
C8H9BrO
201.06
1-(2'Bromophenyl)-1-hydroxyethane, 1-(2-Bromophenyl)ethanol
5411-56-3
100.0
99%
BrC6H4CH(OH)CH3
MFCD00065001

Specifications

2-Bromo-α-methylbenzyl alcohol
98.5
Authentic
1.5590 to 1.5610 (20°C, 589nm)
Clear yellow
≥98.5%
C8H9BrO
201.06
1-(2'Bromophenyl)-1-hydroxyethane, 1-(2-Bromophenyl)ethanol
5411-56-3
100.0
Glass bottle
25g
Liquid
99%
BrC6H4CH(OH)CH3
MFCD00065001
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bromo-a-methylbenzyl alcohol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS