missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-9-fluorenone, 96%, Thermo Scientific Chemicals

Catalog Number p-7037264
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
3096-56-8
C13H7BrO
259.10
MFCD00010789
MTCARZDHUIEYMB-UHFFFAOYSA-N
2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 9h-fluoren-9-one, 2-bromo, 2-bromanylfluoren-9-one, acmc-209hjk, 2-bromo-9h-fluorenone, ksc318o8f
725831
2-bromofluoren-9-one
BrC1=CC=C2C3=CC=CC=C3C(=O)C2=C1
This item is not returnable. View return policy
3096-56-8
C13H7BrO
259.10
MFCD00010789
MTCARZDHUIEYMB-UHFFFAOYSA-N
2-bromo-9-fluorenone, 2-bromo-9h-fluoren-9-one, 9h-fluoren-9-one, 2-bromo, 2-bromanylfluoren-9-one, acmc-209hjk, 2-bromo-9h-fluorenone, ksc318o8f
725831
2-bromofluoren-9-one
BrC1=CC=C2C3=CC=CC=C3C(=O)C2=C1

2-Bromo-9-fluorenone, is used as organic intermediates functional materials. It is mainly used as best liquid crystal material. It is used as an OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-9-fluorenone, is used as organic intermediates functional materials. It is mainly used as best liquid crystal material. It is used as an OLED intermediate.

Solubility
Solubility in other solvents, soluble in acetone.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 146°C to 148°C
Quantity 1 g
Formula Weight 259.1
Percent Purity 96%
Chemical Name or Material 2-Bromo-9-fluorenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'