missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Description
This material is a popular synthetic precursor for OLED materials. Also used to make devices that efficiently emit a deep blue color.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
This material is a popular synthetic precursor for OLED materials. Also used to make devices that efficiently emit a deep blue color.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
This material is a popular synthetic precursor for OLED materials. Also used to make devices that efficiently emit a deep blue color.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 57°C to 62°C |
Quantity | 25 g |
Beilstein | 2524870 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 273.17 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 2-Bromo-9,9-dimethylfluorene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'