missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-methylpyridine, 98%, Thermo Scientific Chemicals
31.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
6-Bromo-2-picoline is a halogenated pyridine derivative used is a building block in the preparation of nitrogen containing heterocyclic compounds. It may used in following synthesis 6,6?-dimethyl-2,2?-bipyridine, 6-methyl-2-pyridyl-2-pyridylmethanone, 2-methyl-6-(trimethylsilyl)-pyridine and N,N’-bis-(6-methylpyrid-2-yl)-(1R,2R)-1,2- diaminocyclohexane (cydiampy).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Bromo-2-picoline is a halogenated pyridine derivative used is a building block in the preparation of nitrogen containing heterocyclic compounds. It may used in following synthesis 6,6′-dimethyl-2,2′-bipyridine, 6-methyl-2-pyridyl-2-pyridylmethanone, 2-methyl-6-(trimethylsilyl)-pyridine and N,N’-bis-(6-methylpyrid-2-yl)-(1R,2R)-1,2- diaminocyclohexane (cydiampy).
Solubility
Soluble in chloroform, ethyl aceate. Not miscible or difficult to mix with water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
6-Bromo-2-picoline is a halogenated pyridine derivative used is a building block in the preparation of nitrogen containing heterocyclic compounds. It may used in following synthesis 6,6′-dimethyl-2,2′-bipyridine, 6-methyl-2-pyridyl-2-pyridylmethanone, 2-methyl-6-(trimethylsilyl)-pyridine and N,N’-bis-(6-methylpyrid-2-yl)-(1R,2R)-1,2- diaminocyclohexane (cydiampy).
Solubility
Soluble in chloroform, ethyl aceate. Not miscible or difficult to mix with water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Density | 1.512 |
Boiling Point | 102°C to 103°C (20 mmHg) |
Flash Point | 97°C (206°F) |
Refractive Index | 1.5625 |
Quantity | 5 g |
Beilstein | 107322 |
Solubility Information | Soluble in chloroform,ethyl aceate. Not miscible or difficult to mix with water. |
Formula Weight | 172.03 |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromo-6-methylpyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'