missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluorobenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number p-7035354
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
913835-80-0
C6H5BBrFO2
218.816
MFCD05664309
MVSHYHSMIRBRGU-UHFFFAOYSA-N
2-bromo-6-fluorophenyl boronic acid, 2-bromo-6-fluorobenzeneboronic acid, 2-bromo-6-fluorophenylboronicacid, boronic acid, b-2-bromo-6-fluorophenyl, acmc-209ra3, 6-bromo-2-fluorophenylboronic acid, 2-bromanyl-6-fluoranyl-phenyl boronic acid
16217456
(2-bromo-6-fluorophenyl)boronic acid
B(C1=C(C=CC=C1Br)F)(O)O
This item is not returnable. View return policy
913835-80-0
C6H5BBrFO2
218.816
MFCD05664309
MVSHYHSMIRBRGU-UHFFFAOYSA-N
2-bromo-6-fluorophenyl boronic acid, 2-bromo-6-fluorobenzeneboronic acid, 2-bromo-6-fluorophenylboronicacid, boronic acid, b-2-bromo-6-fluorophenyl, acmc-209ra3, 6-bromo-2-fluorophenylboronic acid, 2-bromanyl-6-fluoranyl-phenyl boronic acid
16217456
(2-bromo-6-fluorophenyl)boronic acid
B(C1=C(C=CC=C1Br)F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 142°C to 145°C
Quantity 1 g
Formula Weight 218.82
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'