missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-7055963
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
360575-28-6
C7H4BrFO
203.01
MFCD03407341
PJNILWKRAKKEQM-UHFFFAOYSA-N
6-bromo-2-fluorobenzaldehyde, 2-bromo-6-fluorobenzenecarbaldehyde, 2-bromo-6-fluoro-benzaldehyde, 2-fluoro-6-bromobenzaldehyde, 6-fluoro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-6-fluoro, pubchem1419, acmc-1afpf, ksc497k8t
22473977
2-bromo-6-fluorobenzaldehyde
C1=CC(=C(C(=C1)Br)C=O)F
This item is not returnable. View return policy
360575-28-6
C7H4BrFO
203.01
MFCD03407341
PJNILWKRAKKEQM-UHFFFAOYSA-N
6-bromo-2-fluorobenzaldehyde, 2-bromo-6-fluorobenzenecarbaldehyde, 2-bromo-6-fluoro-benzaldehyde, 2-fluoro-6-bromobenzaldehyde, 6-fluoro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-6-fluoro, pubchem1419, acmc-1afpf, ksc497k8t
22473977
2-bromo-6-fluorobenzaldehyde
C1=CC(=C(C(=C1)Br)C=O)F

It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 43°C to 47°C
Density 1.70 g/mL
Boiling Point 100°C to 102°C (8 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2573976
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 203.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-6-fluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'