missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-chloro-4-fluoroaniline, 98%, ACROS Organics™

Brand:  Acros Organics 398440010

Code : 29

Additional Details : CAS Number : 201849-14-1 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III


Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 10376674

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Bromo-6-chloro-4-fluoroaniline
97.5
≥97.5%
C6H4BrClFN
224.46
201849-14-1
100.0
98%
BrC6H2(Cl)(F)NH2
MFCD00051707

Specifications

2-Bromo-6-chloro-4-fluoroaniline
Authentic
100.0
Beige to brown
1g
≥97.5%
C6H4BrClFN
224.46
201849-14-1
97.5
Glass bottle
54° to 58°C
Crystalline Powder
98%
BrC6H2(Cl)(F)NH2
MFCD00051707
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Bromo-6-chloro-4-fluoroaniline

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS