Learn More
Alfa Aesar™ 2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine, 98%
Brand: Alfa Aesar™ H63801.06
Code : NEW
Additional Details : CAS Number : 49669-14-9 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine | |
49669-14-9 | |
244.09 | |
MFCD12406888 |
Specifications
2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine | |
5g | |
98% | |
MFCD12406888 |
49669-14-9 | |
Solid | |
244.09 |