missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-6-(1H-pyrazol-1-yl)pyridine, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 2-bromo-6-1h-pyrazol-1-yl pyridine, 2-bromo-6-pyrazol-1-yl pyridine, 2-bromo-6-pyrazol-1-yl-pyridine, pyridine,2-bromo-6-1h-pyrazol-1-yl, pyridine, 2-bromo-6-1h-pyrazol-1-yl, acmc-209apf, 6-bromo-2-pyridyl pyrazole, 2-bromo-6-1-pyrazolyl pyridine, 2-bromanyl-6-pyrazol-1-yl-pyridine, 1-6-bromo-2-pyridinyl-1h-pyrazole |
Formula Weight | 224.06 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromo-6-(1H-pyrazol-1-yl)pyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'